missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cole-Parmer™ Oakton™ Testr™ Meter Accessories
Replace electrodes when they break, no longer hold calibration, cannot be rehydrated, or are not compatible with your sample.
Supplier: Cole-Parmer™ 3563437
Requires : WD-35634-35
Description
Handy replacement and accessory itemsWarranty
1 Year
Specifications
| ±0.01 | |
| Gray | |
| 3.5 x 1.1 in. | |
| 3.5 x 1.1 x 1.1 in. | |
| Water quality testing, buffer and reagent production, electrochemistry | |
| 3.5 in. | |
| No cable | |
| pH/conductivity/TDS/salinity sensor | |
| pH/conductivity/TDS/salinity sensor | |
| 0 to 10.00 ppt | |
| 0.1 μS, 1 μS; 0.01 mS | |
| 0.1°F/°C | |
| 32.0°F to 140.0°F | |
| 60°C | |
| Yes | |
| Yes | |
| 6 Months | |
| 1.1 in. | |
| None | |
| Oakton Waterproof PCTSTestr 50 |
| ±0.9°F (0.5°C) from 32°F to 122°F (0°C to 50°C); ±1.8°F (1.0°C) from 122°F to 140°F (50°C to 60°C) | |
| Multipin | |
| pH/Conductivity/TDS/Salinity Sensor | |
| Four AAA batteries, installed in meter | |
| Replacement Sensor | |
| 8.89 cm | |
| Valox™ | |
| -1 to 15 | |
| 0 to 20.00 mS | |
| 0 to 10.00 ppt | |
| 0.01 | |
| Yes | |
| 41°F to 113°F | |
| 0°C to 60°C | |
| 5°C to 45°C | |
| 0.2 lb. | |
| 0.09 kg | |
| 2.8 cm | |
| For use with Oakton PCTSTestr™ 50 |