Please login to your online account to display your discounted pricing

PHP 501 trifluoroacetate, Tocris™

Potent GABAA antagonist

$284.81 - $1,180.32

Chemical Identifiers

Chemical Name or Material PHP 501 trifluoroacetate
Assay Percent Range >98%
Molecular Formula C20H21N3O·CF3CO2H
Formula Weight 433.42
Synonym 4-(5-[1, 1'-Biphenyl]-3-yl-1-hydroxy-1H-pyrazol-4-yl)piperidine trifluoroacetate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

446610

 
r & d systems
RDS446610
10mg Each for $284.81

446650

 
r & d systems
RDS446650
50mg Each for $1,180.32
Description & Specifications

Specifications

Formula Weight 433.42
Quantity 10mg
Molecular Formula C20H21N3O·CF3CO2H
Recommended Storage Store at -20°C
Chemical Name or Material PHP 501 trifluoroacetate
Assay Percent Range >98%
CAS 1236105-75-1
Synonym 4-(5-[1, 1'-Biphenyl]-3-yl-1-hydroxy-1H-pyrazol-4-yl)piperidine trifluoroacetate
Safety and Handling

Store at -20°C