missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Photo Numbers and Flags

For 1–9 as numbers or flags.

Manufacturer:  Fisher Science Education 5147

 View more versions of this product

Catalog No. S69030B



Description

Description

Big, bold, and easy to read, these adhesive-backed numbers and flags will adhere to virtually any surface

Photo Numbers

  • Dimensions: 2 × 3 in.
  • Pack of five sets
Photo flags
  • Dimensions: 4 × 5 in.
  • Individual sets of nine

Specifications

Specifications

4 x 5 in.
Adhesive-backed
Education
White
Photo Flag
Documents