Please login to your online account to display your discounted pricing

Photo Manual and Dissection Guides


Use images to prepare students for animal dissections.

Manufacturer: fisher science education  ISBN0-7570-0034-7

 View more versions of this product

Catalog No. S66354

  • / Each
  • / Case of 56 Each
 

Description & Specifications

Specifications

Product Type Photo Manual Dissect Frog
Author Fred Bohensky
Category Biology
For Use With (Application) Study of anatomical structures
Format Wire-O Bound Paperback
Grade High School