Please login to your online account to display your discounted pricing

Photo Manual and Dissection Guides

Use images to prepare students for animal dissections.

$24.30 - $25.40

Specifications

Author Fred Bohensky
Category Biology
Format Paperback
For Use With (Application) Study of anatomical structures
Grade High School
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    

S66354

 
fisher science education
ISBN0-7570-0034-7
Photo Manual Dissect Frog Each for $24.85
Case of 56 Each for $1,390.40

S66353

 
fisher science education
ISBN0-7570-0030-4
Photo Manual and Dissection Guide of the Fetal Pig Each for $25.40
Case of 40 Each for $1,014.85

S66355

 
fisher science education
ISBN0-7570-0033-9
Photo Manual and Dissection Guide of the Rat Each for $24.30
Case of 34 Each for $814.20
Description & Specifications

Specifications

Author Fred Bohensky
Category Biology
Format Paperback
For Use With (Application) Study of anatomical structures
Grade High School

  • Guide students through the dissection process with these easy-to-follow dissection books
  • Lay-flat binding holds book open while preforming the dissection
  • Each chapter concludes with a test for self-study or course evaluation
  • Relates classroom dissections to human life processes with important information on human organs
  • Compatible with most anatomy courses
  • Also includes dissections of up to three sheep organs—heart, eye, or brain