Please login to your online account to display your discounted pricing

Phosphotungstic Acid, n-Hydrate, Crystal, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$519.62 - $1,523.72

Chemical Identifiers

Chemical Name or Material Phosphotungstic Acid
CAS 12067-99-1
Name Note n-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Molecular Formula P2O5·24WO3·nH2O
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003618

 
avantor performance
289101
Poly Bottle 500g Each for $1,523.72

02003619

 
avantor performance
289104
Poly Bottle 125g Each for $519.62
Description & Specifications

Specifications

Physical Form Crystals
Chemical Name or Material Phosphotungstic Acid
Molecular Formula P2O5·24WO3·nH2O
CAS 12067-99-1
Packaging Poly Bottle
Name Note n-Hydrate
Purity Grade BAKER ANALYZED™ Reagent
Specific Gravity 1L = 2.13kg
Quantity 500g