Please login to your online account to display your discounted pricing

Phosphotungstic acid hydrate, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Phosphotungstic acid hydrate
CAS 12067-99-1
Molecular Formula P2O5·24WO3·xH2O
Formula Weight 5706.35 (Anhydrous)
MDL Number MFCD00041964
UN Number UN3260
Merck Index 14,7364
Synonym Tungstophosphoric acid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA4011622

 
alfa aesar
4011622
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 5706.35 (Anhydrous)
Quantity 100g
Chemical Name or Material Phosphotungstic acid hydrate
TSCA TSCA
Physical Form Powder
Molecular Formula P2O5·24WO3·xH2O
MDL Number MFCD00041964
CAS 12067-99-1
UN Number UN3260
Merck Index 14,7364
Synonym Tungstophosphoric acid
Solubility Information Soluble in water, alcohol and ether
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314-H318;
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
Odor Odorless
RTECS Number TH5650000
Color White to Off-white

Applications
Phosphotungstic acid is used as a catalyst for the hydrolysis of propene to prepare 2-propanol. It is also involved in heterogeneous catalysis reaction to prepare propene from 2-propanol and methanol from hydrocarbons. Its ethanolic solution is used in biology as well as to precipitate proteins. It is utilized to precipitate different types of dye such as basic dyes and triphenylmethane dyes. In histology, it is used for staining of cell specimens. It is used in the gravimetric analysis of potassium in analytical chemistry.

Solubility
Soluble in water, alcohol and ether.

Notes
Incompatible with strong bases, strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a;
Do not breathe dust/fume/gas/mist/vapours/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318;
Causes severe skin burns and eye damage.
Causes serious eye damage.