Please login to your online account to display your discounted pricing

Phosphotungstic acid hydrate, ACROS Organics™


Quantity 2.5kg
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  AC208310025

 View more versions of this product

Catalog No. AC208310025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 2880.17
Quantity 2.5kg
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula H3PO4·12WO3·xH2O
Name Note p.a.
Chemical Name or Material Phosphotungstic acid hydrate
Molecular Formula H3O40PW12·xH2O
CAS 12501-23-4
MDL Number MFCD00149915
Packaging Plastic bottle
Synonym Tungstophosphoric acid hydrate
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material Phosphotungstic acid hydrate
Name Note p.a.
CAS 12501-23-4
Molecular Formula H3O40PW12·xH2O
Linear Formula H3PO4·12WO3·xH2O
Formula Weight 2880.17
MDL Number MFCD00149915
Synonym Tungstophosphoric acid hydrate
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents