Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphorylated Cholecystokinin (10-20), Certified Mass Spec Grade

For cellular and molecular biology applications

Specifications

Product Type Phosphorylated Cholecystokinin (10-20), Certified Mass Spec Grade
Molecular Weight 1331.4
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence IKNLQ(pS)LDPSH
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ67710LPL

 
alfa aesar™
J67710LPL
1 vial, 500 pmol CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ67710ODL

 
alfa aesar™
J67710ODL
3 vials, 500 pmol/vial CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Phosphorylated Cholecystokinin (10-20), Certified Mass Spec Grade
Molecular Weight 1331.4
Storage Requirements -30° to -10°C
Format Lyophilized powder
Sequence IKNLQ(pS)LDPSH
Regulatory Status RUO
Purification Method HPLC