missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus(V) oxybromide, 98% min, Alfa Aesar

For Use As a Chemical Intermediate

Manufacturer:  Alfa Aesar 5613514

 View more versions of this product

Catalog No. AA5613514



Specifications

Specifications

Phosphorus(V) tribromide oxide
98% min.
MFCD00036298
UXCDUFKZSUBXGM-UHFFFAOYSA-N
286.685
286.73
2.822g/cm3 at 20°C
25g
7789-59-5
Br3OP
phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3
O=P(Br)(Br)Br
24613
Lump
White to Yellow

Chemical Identifiers

7789-59-5
286.685
UXCDUFKZSUBXGM-UHFFFAOYSA-N
24613
Br3OP
MFCD00036298
phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3
O=P(Br)(Br)Br
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: III

EINECSNumber : 232-177-7

SDS Documents