missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphorus(V) oxychloride, 99.999% (metals basis)

Manufacturer:  Alfa Aesar™ 1184706*

 View more versions of this product

Catalog No. AA1184706



Specifications

Specifications

Phosphorus(V) oxychloride
(metals basis)
POCl3
UN1810
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
O=P(Cl)(Cl)Cl
24813
153.33
99.999%
Pungent
Moisture Sensitive
Colorless
5g
10025-87-3
Cl3OP
MFCD00011443
14,7349
XHXFXVLFKHQFAL-UHFFFAOYSA-N
153.323
CHEBI:30336
Liquid
1.645g/mL
1.460
105.8°C
1.25°C

Chemical Identifiers

10025-87-3
153.323
XHXFXVLFKHQFAL-UHFFFAOYSA-N
24813
O=P(Cl)(Cl)Cl
Cl3OP
MFCD00011443
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
CHEBI:30336
Safety and Handling

Safety and Handling

GHS H Statement
H330-H372-H314-H302
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS OXYCHLORIDE

RTECSNumber : TH4897000

TSCA : TSCA

Documents