missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphorus(V) oxychloride, 99.999% (metals basis)

$58.65 - $58.65

Chemical Identifiers

CAS 10025-87-3
Linear Formula POCl3
Formula Weight 153.33
MDL Number MFCD00011443
UN Number UN1810
Merck Index 14,7349
Synonym Phosphorus(V) trichloride oxide; Phosphoryl chloride
Chemical Name or Material Phosphorus(V) oxychloride
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1184706 SDS Alfa Aesar™
1184706*
5g Each for $58.65
AA1184714 SDS Alfa Aesar™
1184714
25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA1184722 SDS Alfa Aesar™
1184722
100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

10025-87-3
153.33
UN1810
Phosphorus(V) trichloride oxide; Phosphoryl chloride
POCl3
MFCD00011443
14,7349
Phosphorus(V) oxychloride

Specifications

1.645g/mL
Moisture Sensitive
Colorless
5g
1.460
POCl3
MFCD00011443
14,7349
Phosphorus(V) oxychloride
Pungent
105.8°C
1.25°C
10025-87-3
Liquid
153.33
UN1810
Phosphorus(V) trichloride oxide; Phosphoryl chloride
99.999%, (metals basis)

Safety and Handling

GHS H Statement
H330-H372-H314-H302
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Causes severe skin burns and eye damage.
Harmful if swallowed.

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS OXYCHLORIDE

RTECSNumber : TH4897000

TSCA : TSCA