missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus(V) oxychloride, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 010525A1

 View more versions of this product

Catalog No. AA10525A1



Description

Description

Phosphorus(V) oxychloride is widely used as a dehydrating agent and a chlorinating agent. Useful for the conversion of amide to nitriles, Bischler-Napieralski reaction, Vilsmeier Haak reaction, and other reactions producing commercially important intermediates. Acts a source of liquid phosphorus in semiconductor industry. Manufacture of trialkyl phosphate esters and triarylphosphate esters, including commercially popular tricresyl phosphate,which is useful as a plasticizer, flame retardant, gasoline additive, hydrolytic fluid, etc.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

105.8°C
1.25°C
Colorless
1 kg
Cl3OP
MFCD00011443
14,7349
Phosphorus(V) oxychloride is soluble in many organic solvents.
O=P(Cl)(Cl)Cl
24813
153.33
Pungent
Moisture sensitive
36 hPa (27mm Hg) at 20°C
Phosphorus(V) oxychloride
10025-87-3
Liquid
99%
POCl3
UN1810
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
XHXFXVLFKHQFAL-UHFFFAOYSA-N
153.323
CHEBI:30336
99%
1.46
Colorless liquid
1.685 g/cm3 at 20°C

Chemical Identifiers

10025-87-3
153.323
XHXFXVLFKHQFAL-UHFFFAOYSA-N
24813
O=P(Cl)(Cl)Cl
Cl3OP
MFCD00011443
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
CHEBI:30336
Safety and Handling

Safety and Handling

Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P234-P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H330-H335-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS OXYCHLORIDE

EINECSNumber : 233-046-7

RTECSNumber : TH4897000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only