Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphorus(V) oxide, Puratronic™, 99.9925% (metals basis)

Chemical Identifiers

Chemical Name or Material Phosphorus(V) oxide
CAS 1314-56-3
Purity Grade Puratronic™
Formula Weight 141.94
MDL Number MFCD00011440
UN Number UN1807
Merck Index 14,7355
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1082709

  SDS
alfa aesar™
1082709
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1082722

  SDS
alfa aesar™
1082722
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1082759

  SDS
alfa aesar™
1082759
5 x 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Phosphorus(V) oxide
Density 2.39g/mL
Odor Weak
Sensitivity Moisture Sensitive
Boiling Point 360°C (subl.)
Melting Point 340°C
Quantity 10g
CAS 1314-56-3
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H314
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTOXIDE
RTECS Number TH3945000
TSCA TSCA
Purity Grade Puratronic™
Physical Form Powder
Assay 99.9925%
Assay Percent Notes (metals basis)
Formula Weight 141.94
MDL Number MFCD00011440
UN Number UN1807
Merck Index 14,7355
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314
Causes severe skin burns and eye damage.