missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphorus(V) oxide, Puratronic™, 99.9925% (metals basis)

Manufacturer:  Alfa Aesar™ 1082759

 View more versions of this product

Catalog No. AA1082759



Specifications

Specifications

Phosphorus(V) oxide
Weak
360°C (sublimation)
5 x 100g
(metals basis)
UN1807
141.94
99.9925%
2.39g/mL
Moisture Sensitive
340°C
1314-56-3
MFCD00011440
14,7355
Powder
Puratronic™

Chemical Identifiers

1314-56-3
MFCD00011440
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: PHOSPHORUS PENTOXIDE

RTECSNumber : TH3945000

TSCA : TSCA

Documents