missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus(V) oxide, 99.99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 8996609

 View more versions of this product

Catalog No. AA8996609



Specifications

Specifications

Phosphorus(V) oxide
99.99%
MFCD00011440
Powder
Odorless
10g
1314-56-3
P2O5
141.94
2.39g/cm3 at 20°C
White

Chemical Identifiers

1314-56-3
P2O5
MFCD00011440
Safety and Handling

Safety and Handling

H314-H318: Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 215-236-1

SDS Documents