missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus(V) chloride, 98%, Alfa Aesar

For Use As Dehydrating and Chlorinating Agent, Catalyst

Manufacturer:  Thermo Fisher Scientific 1052322

 View more versions of this product

Catalog No. AA1052322



Specifications

Specifications

Phosphorus(V) chloride
98%
MFCD00011439
pcl5
P(Cl)(Cl)(Cl)(Cl)Cl
208.224
CHEBI:30335
Crystals
Pungent
Yellow
10026-13-8
Cl5P
Yellowish crystals
UHZYTMXLRWXGPK-UHFFFAOYSA-N
pentachloro-$l^{5}-phosphane
24819
208.24
2.114g/cm3 at 20°C
0.02 hPa at 20°C
100g

Chemical Identifiers

10026-13-8
208.224
UHZYTMXLRWXGPK-UHFFFAOYSA-N
24819
pentachloro-$l^{5}-phosphane
Cl5P
MFCD00011439
pcl5
CHEBI:30335
P(Cl)(Cl)(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

H330-H302-H314-H373: Fatal if inhaled. Harmful if swallowed. Causes severe skin burns and eye damage. May cause damage to organs through prolonged or repeated exposure.

DOTInformation : DOT Class: 8, Packaging Group: II

EINECSNumber : 233-060-3

storageNote1 : Moisture Sensitive

Documents