missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphorus tribromide, 1.0M solution in dichloromethane, AcroSeal™, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 382068000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentSpecifications
Phosphorus tribromide | |
7789-60-8,75-09-2 | |
Liquid | |
Br3P | |
MFCD00011436 | |
phosphorus tribromide, phosphorous tribromide, tribromophosphine, phosphorus iii bromide, phosphorous bromide, pbr3, phosphorus bromide, phosphorus bromide pbr3, unii-58r3866pua, tribro-mophosphine | |
P(Br)(Br)Br | |
270.69 | |
270.69 | |
AcroSeal™ Glass Bottle | |
1.4880g/mL |
1.0M Solution in Dichloromethane | |
Colorless to Yellow | |
800 mL | |
PBr2 | |
15, 7469 | |
IPNPIHIZVLFAFP-UHFFFAOYSA-N | |
tribromophosphane | |
24614 | |
0.9 to 1.1M | |
1.488 |
RUO – Research Use Only