Please login to your online account to display your discounted pricing

Phosphorus Standard, 1000μg/mL in water with dilute nitric acid, Ultra Scientific

$93.23 - $93.23

Chemical Identifiers

Chemical Name or Material Phosphorus Standard
CAS 007732-18-5
CAS Max % 97.9
CAS 007697-37-2
CAS Max % 2.0
CAS 007722-76-1
CAS Max % 0.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

US-ICP-015

 
ultra scientific
ICP015
125mL Each for $93.23
Description & Specifications

Specifications

Quantity 125mL
Analyte or Component Names Water,Nitric acid,Phosphorus
Solvent or Matrix Water with dilute nitric acid
CAS Max % 97.9
CAS Max % 2.0
CAS Max % 0.1
Concentration or Composition (by Analyte or Components) 1000μg/mL
CAS 007697-37-2
CAS 007732-18-5
Chemical Name or Material Phosphorus Standard
CAS 007722-76-1