missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphorus Standard For ICP, 10,000μg/mL in H2O, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$372.53 - $889.26
Chemical Identifiers
CAS | 7722-76-1 |
---|---|
Molecular Formula | H6NO4P |
Molecular Weight (g/mol) | 115.03 |
MDL Number | MFCD00003396 |
InChI Key | LFVGISIMTYGQHF-UHFFFAOYSA-N |
PubChem CID | 24402 |
ChEBI | CHEBI:62982 |
SMILES | N.OP(O)(O)=O |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
Analyte | Concentration |
---|---|
P | 10,000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
CHEBI:62982 |
H6NO4P | |
MFCD00003396 | |
24402 | |
N.OP(O)(O)=O |
Specifications
7722-76-1,7732-18-5 | |
125 mL | |
Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
H6NO4P | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
phosphoric acid amine | |
24402 | |
Assurance | |
Phosphorus Standard |
Triple-leached LDPE bottle | |
Assurance Certificate of Analysis reports up to 30 elements at ppb levels. | |
AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
MFCD00003396 | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
H2O |
Safety and Handling
Causes skin irritation, serious eye irritation
DOTInformation : Nonhazardous
Recommended Storage : Ambient
Product intended for laboratory use only