missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus, plasma standard solution, Specpure™, P 1000μg/mL, Alfa Aesar™

Manufacturer:  Alfa Aesar 13862AP

 View more versions of this product

Catalog No. AA13862AP

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Phosphorus plasma standard solution
P in 5% HNO3
UN3264
Specpure™
P 1000μg/mL
MFCD00133771
Liquid
500mL

Chemical Identifiers

P in 5% HNO3
MFCD00133771
Safety and Handling

Safety and Handling

GHS H Statement
H314

GHS P Statement
P260-P280-P303+P361+P353-P305+P351+P338-P301+P330+P331-P304+P340-P310-P363-P405-P501a

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents