Please login to your online account to display your discounted pricing

Phosphorus Pentoxide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 125g
Packaging Wide Mouth Amber Glass Bottle

Manufacturer: avantor performance  937404

 View more versions of this product

Catalog No. 02003616

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 141.94
Chemical Name or Material Phosphorus Pentoxide
CAS 1314-56-3
Molecular Formula P2O5
Physical Form Powder
Quantity 125g
Packaging Wide Mouth Amber Glass Bottle
Specific Gravity 1L = 2.39kg

Specifications

Chemical Name or Material Phosphorus Pentoxide
CAS 1314-56-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula P2O5
Formula Weight 141.94