Learn More
Phosphorus pentoxide, 98%, extra pure, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 200890025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentSpecifications
Phosphorus pentoxide | |
1314-56-3 | |
Hygroscopic Crystalline Powder | |
98% | |
P2O5 | |
01,871; 07,291; 10,319; 14,151; 15,267 | |
Phosphoric anhydride | |
DLYUQMMRRRQYAE-UHFFFAOYSA-N | |
tricyclo[3.3.1.1³,â·]tetraphosphoxane-1,3,5,7-tetrone | |
141.94 | |
Extra Pure |
340.0°C to 360.0°C | |
White | |
2.5 kg | |
O5P2 | |
MFCD00011440 | |
15,7467 | |
Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat | |
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 | |
141.94 | |
98% | |
Glass bottle |
Chemical Identifiers
1314-56-3 | |
141.94 | |
DLYUQMMRRRQYAE-UHFFFAOYSA-N | |
tricyclo[3.3.1.1³,â·]tetraphosphoxane-1,3,5,7-tetrone |
O5P2 | |
MFCD00011440 | |
Phosphoric anhydride | |
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3 |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 215-236-1
RUO – Research Use Only