missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus pentoxide, 98+%, ACS reagent, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 315822500

 View more versions of this product

Catalog No. AC315822500



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Specifications

Phosphorus pentoxide
98+%
White
250 g
O5P2
MFCD00011440
01,871; 07,291; 10,319; 14,151; 15,267
Phosphoric anhydride
DLYUQMMRRRQYAE-UHFFFAOYSA-N
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
141.94
ACS Reagent
2.39
340.0°C to 360.0°C
1314-56-3
Powder
98+%
P2O5
1807
15, 7467
Solubility in water: 85% (20°C); reacts forming h3po4 with evol.of heat.
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
141.94
≥98%
Glass bottle

Chemical Identifiers

1314-56-3
141.94
DLYUQMMRRRQYAE-UHFFFAOYSA-N
tricyclo[3.3.1.1³,⁷]tetraphosphoxane-1,3,5,7-tetrone
O5P2
MFCD00011440
Phosphoric anhydride
O=P12OP3(=O)OP(=O)(O1)OP(=O)(O2)O3
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 215-236-1

RTECSNumber : TH3945000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only