Learn More
Phosphorus oxybromide, 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 388450100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
C-X bond formation reagentSpecifications
Phosphorus oxybromide | |
10g | |
Br3OP | |
phosphorus oxybromide, phosphoric tribromide, phosphoryl bromide, phosphoryl tribromide, phosphorusoxybromide, phosphorus v oxybromide, unii-h98h8y87qq, phosphorousoxybromide, phosphoroyl tribromide, pobr3 | |
O=P(Br)(Br)Br | |
24613 | |
95% |
7789-59-5 | |
94% min. (Argentometry) | |
15,746 | |
UXCDUFKZSUBXGM-UHFFFAOYSA-N | |
286.69 | |
286.69 | |
Glass bottle |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 232-177-7
Recommended Storage : 4°C
RUO â Research Use Only