Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphorus(III) fluoride, 99%

Chemical Identifiers

Chemical Name or Material Phosphorus(III) fluoride
CAS 7783-55-3
Formula Weight 87.97
MDL Number MFCD00040325
UN Number UN1955
Merck Index 14,7359
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA3952518

  SDS
alfa aesar™
3952518
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 87.97
Packaging Ampule
Color Colorless
DOT Information Transport Hazard Class: 2.3; Proper Shipping Name: COMPRESSED GASES, TOXIC, N.O.S.
Boiling Point −101.5°C
Physical Form Gas
UN Number UN1955
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,7359
Quantity 50g
Health Hazard 2 GHS H Statement
H330-H314-H318-H302-H312
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
RTECS Number TH3850000
Density 3.907g/mL
Melting Point −151.5°C
Assay 99%
TSCA TSCA
Odor Odorless
MDL Number MFCD00040325
CAS 7783-55-3
Sensitivity Moisture Sensitive
Chemical Name or Material Phosphorus(III) fluoride
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H330-H314-H318-H302-H312
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.