missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphorus(III) chloride, 99.997% (metals basis)

Manufacturer:  Alfa Aesar™ 2298822

 View more versions of this product

Catalog No. AA2298822



Specifications

Specifications

Phosphorus(III) chloride
7719-12-2
Cl3P
UN1809
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
P(Cl)(Cl)Cl
137.32
CHEBI:30334
Liquid
1.574g/mL at 21°C
1.516
−111.8°C
76°C
(metals basis)
MFCD00011438
14,7358
FAIAAWCVCHQXDN-UHFFFAOYSA-N
trichlorophosphane
24387
137.33
99.997%
Acrid
Moisture Sensitive
100g

Chemical Identifiers

7719-12-2
137.32
FAIAAWCVCHQXDN-UHFFFAOYSA-N
24387
trichlorophosphane
Cl3P
MFCD00011438
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
CHEBI:30334
P(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H300-H330-H314-H318-H373
Fatal if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: PHOSPHORUS TRICHLORIDE

TSCA : TSCA

SDS Documents