missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphorus(III) chloride, 98% min., Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4443630

 View more versions of this product

Catalog No. AA4443630



Specifications

Specifications

Phosphorus(III) Chloride
98% min.
MFCD00011438
fosfortrichloride
FAIAAWCVCHQXDN-UHFFFAOYSA-N
trichlorophosphane
24387
137.33
1.574 g/cm3 at 20°C
1.516
76°C
-111.8°C
7719-12-2
Cl3P
1809
Miscible with benzene,carbon disulfide,ether,chloroform and carbon tetrachloride.
P(Cl)(Cl)Cl
137.32
CHEBI:30334
Liquid
Acrid
95mm Hg at 20°C
Yellow
250g

Chemical Identifiers

7719-12-2
137.32
FAIAAWCVCHQXDN-UHFFFAOYSA-N
24387
trichlorophosphane
Cl3P
MFCD00011438
fosfortrichloride
CHEBI:30334
P(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

Keep container tightly closed and dry. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible).

P260-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H300+H330-H314-H373

DOTInformation : DOT Class: 6.1, Packaging Group: I

EINECSNumber : 231-749-3

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents