Please login to your online account to display your discounted pricing

Phosphorus ICP Standard, 1mL = 1mg P (1,000ppm P)NH4H2PO4 in H2O, Ricca Chemical

$156.79 - $307.79

Chemical Identifiers

Chemical Name or Material Phosphorus ICP Standard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7722-76-1
CAS Min % 0.36
CAS Max % 0.38
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

PP1KW-100

 
ricca chemical company
RIPP1KW100
Poly natural 100mL Each for $156.79

PP1KW-500

 
ricca chemical company
RIPP1KW500
Poly natural 500mL Each for $307.79
Description & Specifications

Specifications

Color Clear colorless
Physical Form Liquid
Quantity 100mL
Concentration or Composition (by Analyte or Components) 1mL = 1mg P (1, 000ppm P)NH4H2PO4 in H2O
Chemical Name or Material Phosphorus ICP Standard
Shelf Life 18 months
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7722-76-1
Packaging Poly natural
CAS Min % 0.36
CAS Max % 0.38
Safety and Handling

18 months