Please login to your online account to display your discounted pricing

Phosphorus, AAS Standard Solution, P 1000μg/mL, Alfa Aesar™, Specpure™

$91.77 - $91.77

Chemical Identifiers

Chemical Name or Material Phosphorus
CAS 7697-37-2
CAS 7723-14-0
CAS 7732-18-5
Molecular Formula P in 5% HNO3
MDL Number MFCD00133771
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA89887AE

  SDS
alfa aesar
89887AE
100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA89887AP

  SDS
alfa aesar
89887AP
500mL Each for $91.77
Description & Specifications

Specifications

Quantity 100mL
Color Clear
DOT Information DOT Class: 8, Packaging Group: III
EINECS Number 231-714-2
Appearance Clear liquid
Molecular Formula P in 5% HNO3
MDL Number MFCD00133771
CAS 7697-37-2
Health Hazard 1 H314 : Causes severe skin burns and eye damage.
CAS 7723-14-0
CAS 7732-18-5
Chemical Name or Material Phosphorus
Safety and Handling

H314 : Causes severe skin burns and eye damage.