missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphorus AA Standard, 1000 ppm P in H2O, Ricca Chemical
Supplier: Ricca Chemical Company AP1KW500
Specifications
Phosphorus AA Standard | |
Natural Poly Bottle | |
98.0,0.36 | |
Colorless | |
MFCD00003396 | |
N.OP(O)(O)=O | |
115.03 | |
CHEBI:62982 | |
Liquid |
7722-76-1,7732-18-5 | |
500 mL | |
100.0,0.38 | |
H6NO4P | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
phosphoric acid amine | |
24402 | |
1mL = 1mg P (1, 000ppm P)NH4H2PO4 in H2O |
Chemical Identifiers
7722-76-1 | |
115.03 | |
LFVGISIMTYGQHF-UHFFFAOYSA-N | |
CHEBI:62982 | |
N.OP(O)(O)=O |
H6NO4P | |
MFCD00003396 | |
24402 | |
phosphoric acid amine |
Safety and Handling
ShelfLife : 24 months