Please login to your online account to display your discounted pricing

Phosphoric Acid-Sulfamic Acid Solution, For Chlorine, LabChem™

$81.80 - $81.80

Chemical Identifiers

Chemical Name or Material Phosphoric Acid-Sulfamic Acid Solution
CAS 7732-18-5
CAS Max % 88
CAS 7664-38-2
CAS Max % 10
CAS 5329-14-6
CAS Max % 2
Name Note For Chlorine
Linear Formula H3PO4
Formula Weight 97.1
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC186601

 
lab chem
LC186601
500mL Each for $81.80
Description & Specifications

Specifications

Formula Weight 97.1
Chemical Name or Material Phosphoric Acid-Sulfamic Acid Solution
CAS Max % 2
Decomposition Information Phosphorus oxides; Sulfur compounds
Color Clear, colorless
Quantity 500mL
Name Note For Chlorine
Physical Form Liquid
Linear Formula H3PO4
CAS 7732-18-5
UN Number UN1805
Health Hazard 1 Danger
CAS Max % 88
Health Hazard 2 GHS H Statement
Causes severe skin burns and eye damage.
Identification Passes Test
CAS 7664-38-2
Health Hazard 3 GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
CAS Max % 10
Solubility Soluble in water
CAS 5329-14-6
Storage Information Room Temperature
Safety and Handling

Danger

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Causes severe skin burns and eye damage.