Please login to your online account to display your discounted pricing

Phosphoric Acid NF/FCC, Macron Fine Chemicals™


macron

Quantity 2.5L
Packaging Glass Bottle

Manufacturer: avantor performance  278846

 View more versions of this product

Catalog No. MK2788212

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 98
Purity Grade FCC/NF
Quantity 2.5L
Packaging Glass Bottle
Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
Molecular Formula H3PO4

Specifications

Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
Purity Grade FCC/NF
Molecular Formula H3PO4
Formula Weight 98