Please login to your online account to display your discounted pricing

Phosphoric Acid, N.F. - F.C.C., J.T.Baker™

$158.59 - $1,636.05

Chemical Identifiers

Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
Purity Grade FCC/NF
Molecular Formula H3PO4
Formula Weight 98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003598

 
avantor performance
026203
Glass Bottle 2.5L Case of 6 Each for $1,636.05

02003599

 
avantor performance
026205
Glass Bottle, Single ship 2.5L Each for $1,208.83

02003600

 
avantor performance
026204
Poly Bottle 4L Each for $1,464.92

02003604

 
avantor performance
026201
Glass Bottle, Mall 500mL Each for $158.59

02003605

 
avantor performance
026202
Glass Bottle 500mL Each for $322.96

02003615

 
avantor performance
026208
Poly Drum 19L Each for $894.18
Description & Specifications

Specifications

Purity Grade FCC/NF
Formula Weight 98
Chemical Name or Material Phosphoric Acid
Molecular Formula H3PO4
Specific Gravity 1L = 1.69kg
Quantity 2.5L
CAS 7664-38-2
Packaging Glass Bottle