Please login to your online account to display your discounted pricing

Phosphoric Acid, Multi-Compendial, NF, J.T.Baker™

$478.13 - $2,420.36

Chemical Identifiers

Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
Purity Grade Multi-Compendial/NF
Molecular Formula H3PO4
Formula Weight 98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003610

 
avantor performance
026803
Glass Bottle 2.5L Each for $2,420.36

02003611

 
avantor performance
026805
Glass Bottle, Single ship 2.5L Each for $478.13

02003612

 
avantor performance
026806
Poly Bottle 4L Each for $2,007.90

02003613

 
avantor performance
026800
PVC Coated Glass Bottle 500mL Each for $1,335.49
Description & Specifications

Specifications

Formula Weight 98
Purity Grade Multi-Compendial/NF
Molecular Formula H3PO4
Packaging Glass Bottle
Chemical Name or Material Phosphoric Acid
Specific Gravity 1L = 1.69kg
Quantity 2.5L
CAS 7664-38-2