Please login to your online account to display your discounted pricing

Phosphoric Acid, Multi-Compendial, NF, J.T.Baker™


Quantity 2.5L
Packaging Glass Bottle, Single ship

Manufacturer: avantor performance  026805

 View more versions of this product

Catalog No. 02003611

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Multi-Compendial/NF
Formula Weight 98
CAS 7664-38-2
Molecular Formula H3PO4
Quantity 2.5L
Specific Gravity 1L = 1.69kg
Packaging Glass Bottle, Single ship
Chemical Name or Material Phosphoric Acid

Specifications

Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
Purity Grade Multi-Compendial/NF
Molecular Formula H3PO4
Formula Weight 98