Please login to your online account to display your discounted pricing

Phosphoric acid, J.T.Baker™


High quality chemicals for laboratory and specialized industrial use

Quantity 500mL
Packaging Clear Glass Bottle

Manufacturer: avantor performance  026001

 View more versions of this product

Catalog No. 02003602

  • / Each
 

Description & Specifications

Chemical Identifiers

Purity Grade Baker Analyzed™ ACS Reagent
Formula Weight 97.994
Chemical Name or Material Phosphoric Acid
Synonym Orthophosphoric Acid; Phosphoric(V) acid
CAS 7664-38-2
Density 1.885g/mL
Color White or Colorless
Quantity 500mL
Specific Gravity 1L = 1.69kg
Melting Point 42.35°C
Boiling Point 158°C
Physical Form Solid or Viscous Liquid
Molecular Formula H3PO4
Packaging Clear Glass Bottle

Specifications

Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula H3PO4
Formula Weight 97.994
Synonym Orthophosphoric Acid; Phosphoric(V) acid

    For Laboratory,Research,or Manufacturing Use