Learn More
Phosphoric acid, extra pure, 85% solution in water, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 295700100
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Phosphoric acid | |
7664-38-2,7732-18-5 | |
10ppm max. | |
1.68 | |
H3O4P | |
MFCD00011340 | |
15, 7456 | |
Solubility in water: miscible. | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 | |
84.5 to 86.0% |
85% Solution in Water | |
1.6800g/mL | |
Plastic drum | |
10L | |
H3PO4 | |
01,860; 04,387; 07,255; 10,317; 15,266; 16,177 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
98 | |
Extra Pure |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Danger
RUO â Research Use Only