Please login to your online account to display your discounted pricing

Phosphoric acid, ACS reagent, 85+% solution in water, ACROS Organics™


Quantity 10L
Packaging Plastic Jerrycan

Manufacturer: thermo fisher scientific  AC424040100

 View more versions of this product

Catalog No. AC424040100

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 98
Purity Grade ACS Reagent Grade
Assay ≥85.0%
Quantity 10L
Color Clear
Merck Index 15, 7456
Packaging Plastic Jerrycan
Density 1.6800g/mL
Fieser 01,860; 04,387; 07,255; 10,317; 15,266; 16,177
Calcium (Ca) 20ppm max.
Sodium (Na) 250ppm max.
Arsenic (As) 1ppm max.
Sulfate 30ppm max.
CAS 7732-18-5
Potassium (K) 50ppm max.
Antimony (Sb) 20ppm max.
Molecular Formula H3O4P
Volatile Acids 0.001% max.
CAS 7664-38-2
MDL Number MFCD00011340
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Iron (Fe) 30ppm max.
Nitrate 5ppm max.
Heavy Metals (as Pb) 10ppm max.
Insoluble Matter 0.001% max.
Synonym Orthophosphoric acid
Additional Information Vapor Pressure: 2hPa at 20°C
Magnesium (Mg) 20ppm max.
Reducing Substances Passes Test
Chlorine (Cl) 3ppm max.
Linear Formula H3PO4
Manganese (Mn) 0.5ppm max.
Name Note 85+% Solution in Water
Chemical Name or Material Phosphoric acid
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Phosphoric acid
Name Note 85+% Solution in Water
CAS 7664-38-2
CAS 7732-18-5
Purity Grade ACS Reagent Grade
Assay ≥85.0%
Molecular Formula H3O4P
Linear Formula H3PO4
Formula Weight 98
MDL Number MFCD00011340
Synonym Orthophosphoric acid
Accessories

Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.

SDS & Safety Documents