missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphoric Acid, ACS Grade, ≥ 85.0% (w/w), LabChem™

Manufacturer:  LabChem LC186403S

 View more versions of this product

Catalog No. LC186403S



Specifications

Specifications

Phosphoric Acid
2.5L
100
H3PO4
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
1,685kg/m3
phosphoric acid
1004
97.1
≥85.0% (w/w)
Passes Test
Passes Test
1685kg/m3
Phosphorus oxides; Carbon monoxide; Carbon dioxide; Thermal decomposition generates: Corrosive vapors
158°C
Colorless
7664-38-2
H3O4P
UN1805
NBIIXXVUZAFLBC-UHFFFAOYSA-N
OP(=O)(O)O
97.994
CHEBI:26078
Liquid
ACS
Poly Bottle
Soluble in water
Room Temperature
21°C

Chemical Identifiers

7664-38-2
97.994
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
CHEBI:26078
OP(=O)(O)O
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/doctor.
If on skin (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Immediately call a poison center/doctor.
Wash contaminated clothing before reuse.
If inhaled: Remove person to fresh air and keep comfortable for breathing.
Immediately call a poison center/doctor.
If in eyes: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

Danger

EINECSNumber : 231-633-2

Documents
Promotions

Promotions