Please login to your online account to display your discounted pricing

Phosphoric Acid ACS AR, Macron Fine Chemicals™

macron

Chemical Identifiers

Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
Purity Grade ACS Grade
Assay Percent Range 85%
Molecular Formula H3PO4
Formula Weight 98
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MK2816212

 
avantor performance
281606
Poly 2.5kg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

MK2816500

 
avantor performance
281604
POLYSTORMOR 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Purity Grade ACS Grade
Formula Weight 98
Quantity 2.5kg
Molecular Formula H3PO4
CAS 7664-38-2
Packaging Poly
Chemical Name or Material Phosphoric Acid
Assay Percent Range 85%