Please login to your online account to display your discounted pricing

Phosphoric Acid, 85%, AR Grade, For ACS analysis, Macron Fine Chemicals™


macron

Quantity 500mL

Manufacturer: avantor performance  279616

 View more versions of this product

Catalog No. MK279616

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 500mL
Formula Weight 98
Purity Grade AR Grade
Physical Form Liquid
Antimony (Sb) max. 15ppm
Arsenic (As) max. 1ppm
Volatile Acids max. 0.001%
Reducing Substances max. 1.0
Color max. 10
Chloride max. 3ppm
Iron (Fe) max. 6ppm
Potassium (K) max. 6ppm
Molecular Formula H3PO4
Heavy Metals max. 10ppm
Calcium (Ca) max. 0.002%
Assay Percent Range min. 85.0%
Insoluble Matter max. 0.001%
Magnesium (Mg) max. 0.002%
CAS 7664-38-2
Purity Grade Notes For ACS analysis (by acidimetry)
Appearance Clear, colorless liquid
Sulfate max. 0.001%
Manganese (Mn) max. 0.5ppm
Nitrate max. 5ppm
Sodium (Na) max. 10ppm

Specifications

CAS 7664-38-2
Purity Grade AR Grade
Purity Grade Notes For ACS analysis (by acidimetry)
Physical Form Liquid
Assay Percent Range min. 85.0%
Molecular Formula H3PO4
Formula Weight 98