Please login to your online account to display your discounted pricing

Phosphoric Acid 85% Acid, ACS Reagent Grade, Ricca Chemical


Quantity 500mL

Manufacturer: ricca chemical company  RIRABP0030500D1

 View more versions of this product

Catalog No. RABP0030500

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 500mL
Purity Grade ACS Reagent Grade
Physical Form Liquid
Color Clear colorless
Concentration or Composition Notes 85% Acid
Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
CAS Min % 85.0
CAS Max % 100.0
CAS 7732-18-5 (Water, Deionized)

Specifications

Chemical Name or Material Phosphoric Acid
CAS 7664-38-2
CAS Min % 85.0
CAS Max % 100.0
CAS 7732-18-5 (Water, Deionized)
Purity Grade ACS Reagent Grade
Physical Form Liquid