missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Phosphoric Acid, 20% (v/v), Ricca Chemical
Supplier: Ricca Chemical Company R585030020F
Specifications
Phosphoric Acid | |
7664-38-2,7732-18-5 | |
81.0,21.0 | |
20 L | |
Liquid | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
phosphoric acid | |
1004 | |
20% (v/v) Aqueous Solution (1 + 4) |
Colorless | |
79.0,19.0 | |
Cubitainer | |
H3O4P | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
OP(=O)(O)O | |
97.994 | |
CHEBI:26078 |
Chemical Identifiers
7664-38-2 | |
97.994 | |
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico | |
CHEBI:26078 | |
OP(=O)(O)O |
H3O4P | |
NBIIXXVUZAFLBC-UHFFFAOYSA-N | |
1004 | |
phosphoric acid |
Safety and Handling
ShelfLife : 24 months