Please login to your online account to display your discounted pricing

Phosphoric Acid, 10% (v/v) Aqueous Solution, Ricca Chemical

$563.24 - $563.24

Chemical Identifiers

Chemical Name or Material Phosphoric Acid
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 91.0
CAS 7664-38-2
CAS Min % 9.0
CAS Max % 11.0
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

5850-25

 
ricca chemical company
RI58502.5
Cubitainer 10L Each for $563.24
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 10L
Chemical Name or Material Phosphoric Acid
Concentration or Composition Notes 10% (v/v)
Solvent or Matrix Aqueous Solution
CAS 7732-18-5
CAS Min % 89.0
CAS Max % 91.0
CAS 7664-38-2
CAS Min % 9.0
CAS Max % 11.0
Packaging Cubitainer