missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphoric Acid, Certified, 10.0% (v/v) ±0.2% (v/v), LabChem™

Manufacturer:  LabChem LC186501

 View more versions of this product

Catalog No. LC186501



Specifications

Specifications

Phosphoric Acid, (1+9)
500mL
90,10
H3PO4
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
OP(=O)(O)O
97.994
CHEBI:26078
Liquid
Certified
Poly Bottle
Room Temperature
Phosphorus oxides; Thermal decomposition generates: Corrosive vapors
Colorless
7664-38-2,7732-18-5
H3O4P
UN1805
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
1004
97.1
10.0% ± 0.2% (v/v)
Passes Test
Soluble in water
Traceable to NIST

Chemical Identifiers

7664-38-2
97.994
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
CHEBI:26078
OP(=O)(O)O
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.

GHS P Statement
Do not breathe mist, vapors, spray.
Wear protective gloves/eye protection.
Wash exposed skin thoroughly after handling.
If swallowed: Rinse mouth.
Do not induce vomiting.
Immediately call a poison center/docto

Danger

Documents
Promotions

Promotions