Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphoramidon disodium salt, ≥97%

Inhibitor of thermolysin and collagenase

Chemical Identifiers

Chemical Name or Material Phosphoramidon disodium salt
CAS 119942-99-3
Formula Weight 587.47
MDL Number MFCD00077870
Beilstein 8105669
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63456LB0

  SDS
alfa aesar™
J63456LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 587.47
CAS 119942-99-3
Physical Form Solid
Assay ≥97%
MDL Number MFCD00077870
Beilstein 8105669
Quantity 5mg
Storage Note 1 -30° to -10°C
Color White to pale yellow
Chemical Name or Material Phosphoramidon disodium salt
Safety and Handling

-30° to -10°C