Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphopeptide Certified Mass Spec Standard II


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67638ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67638ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Format Lyophilized powder
Product Type Phosphopeptide Certified Mass Spec Standard II, 500pm each peptide per vial
Quantity 3 vials, 500 pmol/vial
Regulatory Status RUO
Storage Requirements -30° to -10°C
Purification Method HPLC