Please login to your online account to display your discounted pricing

Alfa Aesar™ Phosphopeptide Certified Mass Spec Standard I


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67696LPL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67696LPL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Quantity 1 vial, 500 pmol
Purification Method HPLC
Sequence Angiotensin II, phosphorylated, Cholecystokinin (10-20), phosphorylated, Calcitonin (15-29), phosphorylated
Product Type Phosphopeptide Certified Mass Spec Standard I, 500pmol each peptide per vial 1 vial
Regulatory Status RUO
Storage Requirements -30° to -10°C
Format Lyophilized powder