missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Phosphonoacetic acid, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC258290250

 View more versions of this product

Catalog No. AC258290250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Phosphonoacetic acid
25g
Authentic
Powder
98%
HO2CCH2P(O)(OH)2
MFCD00004311
White
4408-78-0
Glass bottle
≥97.5%
C2H5O5P
140.03

Chemical Identifiers

98%
HO2CCH2P(O)(OH)2
MFCD00004311
≥97.5%
C2H5O5P
140.03
Phosphonoacetic acid

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger